5-Chloro-2-methyl-3-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

78222-1G 78222-500MG 78222-5G
76935-974EA 1043.93 USD
76935-974 76935-976 76935-978
5-Chloro-2-methyl-3-(methylthio)benzaldehyde ≥97%
5-Chloro-2-methyl-3-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉ClOS
MW: 200.685 g/mol
Storage Temperature: Refrigerator
CAS Number: 2918912-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR