4-Chloro-1-ethyl-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

78189-1G 78189-25G 78189-500MG 78189-5G
76935-960EA 147.47 USD
76935-960 76935-962 76935-964 76935-966
4-Chloro-1-ethyl-1H-pyrazole ≥97%
4-Chloro-1-ethyl-1H-pyrazole
Formula: C₅H₇ClN₂
Storage Temperature: Ambient
MDL Number: MFCD12026905
CAS Number: 1171076-88-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR