5-Bromo-2-(methylthio)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

76661-1G 76661-25G 76661-500MG 76661-5G
76935-948EA 290.69 USD
76935-948 76935-950 76935-952 76935-954
5-Bromo-2-(methylthio)benzonitrile ≥97%
5-Bromo-2-(methylthio)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₆BrNS
Storage Temperature: Ambient
CAS Number: 391200-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR