5-Bromo-2-hydroxy-3-methylbenzenecarboxylic acid ≥95% research grade
Supplier: AOBChem USA
|
Warning
|
Synonyms:
5-Bromo-2-hydroxy-3-methylbenzoic acid
75734-25G
75734-5G
76935-936EA
272.66
USD
76935-936
76935-938
5-Bromo-2-hydroxy-3-methylbenzenecarboxylic acid ≥95% research grade
5-Bromo-2-hydroxy-3-methylbenzenecarboxylic acid
Formula:
C₈H₇BrO₃ MW: 231.05 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00093269 CAS Number: 36194-82-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...