5-Fluoro-2-methoxybenzoyl chloride ≥97%

Supplier: AOBChem USA

Warning

75423-1G 75423-25G 75423-5G
76935-908EA 66.84 USD
76935-908 76935-910 76935-912
5-Fluoro-2-methoxybenzoyl chloride ≥97%
5-Fluoro-2-methoxybenzoyl chloride

Precaución: for Research Use Only

Formula: C₈H₆ClFO₂
MW: 188.583 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09832309
CAS Number: 704-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR