5-Bromo-7-(trifluoromethyl)-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

74793-1G 74793-25G 74793-5G
76935-896EA 106.09 USD
76935-896 76935-898 76935-900
5-Bromo-7-(trifluoromethyl)-1H-indazole ≥97%
5-Bromo-7-(trifluoromethyl)-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₄BrF₃N₂
MW: 265.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21602947
CAS Number: 1374258-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR