4-Bromo-1,3-benzodioxole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromobenzo[d][1,3]dioxole

73729-1G 73729-25G 73729-5G
76935-868EA 78.51 USD
76935-868 76935-870 76935-872
4-Bromo-1,3-benzodioxole ≥97%
4-Bromo-1,3-benzodioxole

Precaución: for Research Use Only

Formula: C₇H₅BrO₂
MW: 201.0174 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02681890
CAS Number: 6698-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR