5-Bromo-2-(p-tolyloxy)pyrimidine ≥97%

Supplier: AOBChem USA

Warning

72964-1G 72964-25G 72964-500MG 72964-5G
76935-854EA 118.83 USD
76935-854 76935-856 76935-858 76935-860
5-Bromo-2-(p-tolyloxy)pyrimidine ≥97%
5-Bromo-2-(p-tolyloxy)pyrimidine

Precaución: for Research Use Only

Formula: C₁₁H₉BrN₂O
MW: 265.106 g/mol
Storage Temperature: Ambient
CAS Number: 887430-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR