4-Iodo-3-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

72642-1G 72642-25G 72642-500MG 72642-5G
76935-838EA 170.81 USD
76935-838 76935-840 76935-842 76935-844
4-Iodo-3-methylbenzamide ≥97%
4-Iodo-3-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₈INO
Storage Temperature: Ambient
CAS Number: 933672-34-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR