5-Ethyl-2-fluoro-3-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

69753-1G 69753-500MG 69753-5G
76935-810EA 1327.19 USD
76935-810 76935-812 76935-814
5-Ethyl-2-fluoro-3-methylbenzaldehyde ≥97%
5-Ethyl-2-fluoro-3-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO
Storage Temperature: Refrigerator
CAS Number: 2710560-72-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR