5-Bromo-4-chlorobenzo[d][1,3]dioxole ≥95%

Supplier: AOBChem USA

Warning

68847-1G 68847-25G 68847-500MG 68847-5G
76935-796EA 411.63 USD
76935-796 76935-798 76935-800 76935-802
5-Bromo-4-chlorobenzo[d][1,3]dioxole ≥95%
5-Bromo-4-chlorobenzo[d][1,3]dioxole

Precaución: for Research Use Only

Formula: C₇H₄BrClO₂
Storage Temperature: Ambient
CAS Number: 72744-46-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR