5-(5-Chloro-2-isopropoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

68112-1G 68112-500MG 68112-5G
76935-766EA 464.68 USD
76935-766 76935-768 76935-770
5-(5-Chloro-2-isopropoxyphenyl)thiazol-2-amine ≥97%
5-(5-Chloro-2-isopropoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₃ClN₂OS
Storage Temperature: Ambient
CAS Number: 2807450-16-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR