4-Bromo-3-methoxy-2-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

66488-1G 66488-500MG 66488-5G
76935-740EA 919.81 USD
76935-740 76935-742 76935-744
4-Bromo-3-methoxy-2-methylbenzoic acid ≥97%
4-Bromo-3-methoxy-2-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
MW: 245.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16658634
CAS Number: 1154060-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR