4-Formyl-2-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

65313-1G 65313-500MG 65313-5G
76935-716EA 584.56 USD
76935-716 76935-718 76935-720
4-Formyl-2-methoxybenzoic acid ≥97%
4-Formyl-2-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₈O₄
MW: 180.157 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20640467
CAS Number: 194928-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR