5-(2-Fluoro-6-isopropoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

65282-1G 65282-500MG 65282-5G
76935-704EA 589.87 USD
76935-704 76935-706 76935-708
5-(2-Fluoro-6-isopropoxyphenyl)thiazol-2-amine ≥97%
5-(2-Fluoro-6-isopropoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₃FN₂OS
MW: 252.3078 g/mol
Storage Temperature: Ambient
CAS Number: 2807470-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR