5-(5-Bromo-2,4-difluorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

65185-1G 65185-500MG 65185-5G
76935-690EA 500.75 USD
76935-690 76935-692 76935-694
5-(5-Bromo-2,4-difluorophenyl)thiazol-2-amine ≥97%
5-(5-Bromo-2,4-difluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₅BrF₂N₂S
MW: 291.1152 g/mol
Storage Temperature: Ambient
CAS Number: 2807464-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR