5-Bromo-3-methyl-2-(2,2,2-trifluoroethoxy)pyridine ≥97%

Supplier: AOBChem USA

Warning

64962-1G 64962-25G 64962-500MG 64962-5G
76935-674EA 203.7 USD
76935-674 76935-676 76935-678 76935-680
5-Bromo-3-methyl-2-(2,2,2-trifluoroethoxy)pyridine ≥97%
5-Bromo-3-methyl-2-(2,2,2-trifluoroethoxy)pyridine

Precaución: for Research Use Only

Formula: C₈H₇BrF₃NO
MW: 270.0464 g/mol
Storage Temperature: Ambient
CAS Number: 1187930-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR