5-Bromo-1-fluoro-2-iodo-3-methylbenzene ≥97%
Supplier: AOBChem USA
|
Warning
|
64670-1G
64670-25G
64670-500MG
64670-5G
76935-628EA
193.09
USD
76935-628
76935-630
76935-632
76935-634
5-Bromo-1-fluoro-2-iodo-3-methylbenzene ≥97%
5-Bromo-1-fluoro-2-iodo-3-methylbenzene
Formula:
C₇H₅BrFI Storage Temperature: Ambient |
MDL Number:
MFCD28739626 CAS Number: 1805553-01-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...