4-Ethoxy-2-fluoro-1-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

64508-1G 64508-25G 64508-5G
76935-618EA 88.06 USD
76935-618 76935-620 76935-622
4-Ethoxy-2-fluoro-1-iodobenzene ≥97%
4-Ethoxy-2-fluoro-1-iodobenzene

Precaución: for Research Use Only

Formula: C₈H₈FIO
MW: 266.0513 g/mol
Storage Temperature: Refrigerator
CAS Number: 107956-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR