5-(Benzyloxy)-2-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

63787-1G 63787-25G 63787-500MG 63787-5G
76935-586EA 359.65 USD
76935-586 76935-588 76935-590 76935-592
5-(Benzyloxy)-2-methoxybenzoic acid ≥97%
5-(Benzyloxy)-2-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₄
Storage Temperature: Refrigerator
CAS Number: 84923-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR