5-(2-Fluoro-4-isopropoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

63374-1G 63374-500MG 63374-5G
76935-576EA 666.25 USD
76935-576 76935-578 76935-580
5-(2-Fluoro-4-isopropoxyphenyl)thiazol-2-amine ≥97%
5-(2-Fluoro-4-isopropoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₃FN₂OS
MW: 252.3078 g/mol
Storage Temperature: Ambient
CAS Number: 2807454-71-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR