5-(2-Fluoro-4-propoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

63363-1G 63363-25G 63363-500MG 63363-5G
76935-546EA 405.27 USD
76935-546 76935-548 76935-550 76935-552
5-(2-Fluoro-4-propoxyphenyl)thiazol-2-amine ≥97%
5-(2-Fluoro-4-propoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₃FN₂OS
MW: 252.3078 g/mol
Storage Temperature: Ambient
CAS Number: 2807445-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR