4-Bromo-2-methoxy-6-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

63209-1G 63209-25G 63209-500MG 63209-5G
76935-530EA 320.4 USD
76935-530 76935-532 76935-534 76935-536
4-Bromo-2-methoxy-6-methylbenzaldehyde ≥97%
4-Bromo-2-methoxy-6-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
Storage Temperature: Refrigerator
CAS Number: 1244949-43-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR