5-Bromo-4-fluoro-2-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

63100-1G 63100-25G 63100-5G
76935-520EA 88.06 USD
76935-520 76935-522 76935-524
5-Bromo-4-fluoro-2-methylbenzaldehyde ≥97%
5-Bromo-4-fluoro-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆BrFO
Storage Temperature: Ambient
CAS Number: 1782824-13-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR