5-Chloro-2-fluoro-4-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

63089-1G 63089-25G 63089-500MG 63089-5G
76935-504EA 206.88 USD
76935-504 76935-506 76935-508 76935-510
5-Chloro-2-fluoro-4-methylbenzaldehyde ≥97%
5-Chloro-2-fluoro-4-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆ClFO
MW: 172.584 g/mol
Storage Temperature: Ambient
CAS Number: 1785064-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR