5-(3-Chloro-2,6-difluorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

62693-1G 62693-25G 62693-500MG 62693-5G
76935-484EA 376.62 USD
76935-484 76935-486 76935-488 76935-490
5-(3-Chloro-2,6-difluorophenyl)thiazol-2-amine ≥97%
5-(3-Chloro-2,6-difluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₅ClF₂N₂S
Storage Temperature: Ambient
CAS Number: 2764731-73-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR