5-(3-Methoxy-5-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

62691-1G 62691-25G 62691-500MG 62691-5G
76935-470EA 296 USD
76935-470 76935-472 76935-474 76935-476
5-(3-Methoxy-5-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%
5-(3-Methoxy-5-(trifluoromethyl)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₉F₃N₂OS
Storage Temperature: Ambient
CAS Number: 2764732-16-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR