5-(2-Bromo-4,5-difluorophenyl)thiazol-2-amine ≥95%

Supplier: AOBChem USA

Warning

62538-1G 62538-500MG 62538-5G
76935-458EA 494.38 USD
76935-458 76935-460 76935-462
5-(2-Bromo-4,5-difluorophenyl)thiazol-2-amine ≥95%
5-(2-Bromo-4,5-difluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₅BrF₂N₂S
MW: 291.1152 g/mol
Storage Temperature: Ambient
CAS Number: 2764733-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR