5-(4-Chloro-3,5-difluorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

62536-1G 62536-500MG 62536-5G
76935-446EA 554.86 USD
76935-446 76935-448 76935-450
5-(4-Chloro-3,5-difluorophenyl)thiazol-2-amine ≥97%
5-(4-Chloro-3,5-difluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₅ClF₂N₂S
MW: 246.6642 g/mol
Storage Temperature: Ambient
CAS Number: 2764728-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR