5-(4-(Benzyloxy)-3-methylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

62528-1G 62528-500MG 62528-5G
76935-408EA 629.12 USD
76935-408 76935-410 76935-412
5-(4-(Benzyloxy)-3-methylphenyl)thiazol-2-amine ≥97%
5-(4-(Benzyloxy)-3-methylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₇H₁₆N₂OS
MW: 296.3867 g/mol
Storage Temperature: Ambient
CAS Number: 2764729-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR