5-(3-Bromo-6-chloro-2-fluorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

62525-1G 62525-500MG 62525-5G
76935-396EA 527.27 USD
76935-396 76935-398 76935-400
5-(3-Bromo-6-chloro-2-fluorophenyl)thiazol-2-amine ≥97%
5-(3-Bromo-6-chloro-2-fluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₅BrClFN₂S
Storage Temperature: Ambient
CAS Number: 2764729-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR