4-Bromo-2-fluoro-3-methylaniline ≥97%

Supplier: AOBChem USA

Warning

62148-1G 62148-25G 62148-500MG 62148-5G
76935-386EA 266.29 USD
76935-386 76935-388 76935-390 76935-392
4-Bromo-2-fluoro-3-methylaniline ≥97%
4-Bromo-2-fluoro-3-methylaniline

Precaución: for Research Use Only

Formula: C₇H₇BrFN
Storage Temperature: Ambient
MDL Number: MFCD26687842
CAS Number: 1540204-53-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR