5-(3-Bromo-4-butoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

61988-1G 61988-500MG 61988-5G
76935-374EA 593.05 USD
76935-374 76935-376 76935-378
5-(3-Bromo-4-butoxyphenyl)thiazol-2-amine ≥97%
5-(3-Bromo-4-butoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₃H₁₅BrN₂OS
MW: 327.24 g/mol
Storage Temperature: Ambient
CAS Number: 2764731-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR