5-(2-Chloro-6-fluoro-3-methylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

61986-1G 61986-500MG 61986-5G
76935-362EA 493.32 USD
76935-362 76935-364 76935-366
5-(2-Chloro-6-fluoro-3-methylphenyl)thiazol-2-amine ≥97%
5-(2-Chloro-6-fluoro-3-methylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₈ClFN₂S
MW: 242.7003 g/mol
Storage Temperature: Ambient
CAS Number: 2764730-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR