4-Bromo-N-ethyl-3-methylbenzamide ≥95%

Supplier: AOBChem USA

Warning

61243-1G 61243-25G 61243-5G
76935-322EA 64.72 USD
76935-322 76935-324 76935-326
4-Bromo-N-ethyl-3-methylbenzamide ≥95%
4-Bromo-N-ethyl-3-methylbenzamide

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrNO
Storage Temperature: Ambient
MDL Number: MFCD09972131
CAS Number: 1020252-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR