5-(2-Bromo-6-fluoro-4-methylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

61141-1G 61141-25G 61141-500MG 61141-5G
76935-296EA 381.93 USD
76935-296 76935-298 76935-300 76935-302
5-(2-Bromo-6-fluoro-4-methylphenyl)thiazol-2-amine ≥97%
5-(2-Bromo-6-fluoro-4-methylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₈BrFN₂S
MW: 287.1513 g/mol
Storage Temperature: Ambient
CAS Number: 2757729-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR