5-(2-Fluoro-4-(methylthio)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

61139-1G 61139-500MG 61139-5G
76935-282EA 558.04 USD
76935-282 76935-284 76935-286
5-(2-Fluoro-4-(methylthio)phenyl)thiazol-2-amine ≥97%
5-(2-Fluoro-4-(methylthio)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₉FN₂S₂
MW: 240.3202 g/mol
Storage Temperature: Ambient
CAS Number: 2764729-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR