5-(3-Isopropyl-4-methoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

61123-1G 61123-500MG 61123-5G
76935-218EA 700.2 USD
76935-218 76935-220 76935-222
5-(3-Isopropyl-4-methoxyphenyl)thiazol-2-amine ≥97%
5-(3-Isopropyl-4-methoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₃H₁₆N₂OS
MW: 248.3439 g/mol
Storage Temperature: Ambient
CAS Number: 2764733-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR