5-(4-Bromo-2-isopropylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

61120-1G 61120-500MG 61120-5G
76935-206EA 702.32 USD
76935-206 76935-208 76935-210
5-(4-Bromo-2-isopropylphenyl)thiazol-2-amine ≥97%
5-(4-Bromo-2-isopropylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₃BrN₂S
MW: 297.214 g/mol
Storage Temperature: Ambient
CAS Number: 2764728-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR