5-Chloro-2,3,4-trifluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

61085-1G 61085-25G 61085-5G
76935-194EA 114.58 USD
76935-194 76935-196 76935-198
5-Chloro-2,3,4-trifluorobenzoic acid ≥97%
5-Chloro-2,3,4-trifluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₂ClF₃O₂
Storage Temperature: Ambient
CAS Number: 101513-73-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR