5-(2-Chloro-3,6-difluorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

60976-1G 60976-25G 60976-500MG 60976-5G
76935-164EA 352.22 USD
76935-164 76935-166 76935-168 76935-170
5-(2-Chloro-3,6-difluorophenyl)thiazol-2-amine ≥97%
5-(2-Chloro-3,6-difluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₅ClF₂N₂S
Storage Temperature: Ambient
CAS Number: 2275829-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR