5-(3-Chloro-6-fluoro-2-methoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

60974-1G 60974-500MG 60974-5G
76935-152EA 495.45 USD
76935-152 76935-154 76935-156
5-(3-Chloro-6-fluoro-2-methoxyphenyl)thiazol-2-amine ≥97%
5-(3-Chloro-6-fluoro-2-methoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₈ClFN₂OS
MW: 258.6997 g/mol
Storage Temperature: Ambient
CAS Number: 2757730-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR