5-(3-Chloro-2-fluoro-5-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

60972-1G 60972-25G 60972-500MG 60972-5G
76935-138EA 369.2 USD
76935-138 76935-140 76935-142 76935-144
5-(3-Chloro-2-fluoro-5-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%
5-(3-Chloro-2-fluoro-5-(trifluoromethyl)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₅ClF₄N₂S
MW: 296.6717 g/mol
Storage Temperature: Ambient
CAS Number: 2764729-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR