5-(2-Fluoro-5-isopropoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

60270-1G 60270-500MG 60270-5G
76935-088EA 672.62 USD
76935-088 76935-090 76935-092
5-(2-Fluoro-5-isopropoxyphenyl)thiazol-2-amine ≥97%
5-(2-Fluoro-5-isopropoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₃FN₂OS
MW: 252.308 g/mol
Storage Temperature: Ambient
CAS Number: 2755718-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR