4-Iodo-1-methyl-1H-pyrazole-5-carbaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
60155-1G
60155-25G
60155-500MG
60155-5G
76935-072EA
135.8
USD
76935-072
76935-074
76935-076
76935-078
4-Iodo-1-methyl-1H-pyrazole-5-carbaldehyde ≥97%
4-Iodo-1-methyl-1H-pyrazole-5-carbaldehyde
Formula:
C₅H₅IN₂O MW: 236.01 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22200307 CAS Number: 959986-66-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...