4-Ethoxy-5-methoxy-2-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

60027-1G 60027-500MG 60027-5G
76935-058EA 560.16 USD
76935-058 76935-060 76935-062
4-Ethoxy-5-methoxy-2-(trifluoromethyl)benzoic acid ≥97%
4-Ethoxy-5-methoxy-2-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₁F₃O₄
Storage Temperature: Ambient
CAS Number: 2702626-31-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR