4-Bromo-2-ethoxy-1-(trifluoromethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

59904-1G 59904-25G 59904-500MG 59904-5G
76935-032EA 129.43 USD
76935-032 76935-034 76935-036 76935-038
4-Bromo-2-ethoxy-1-(trifluoromethoxy)benzene ≥97%
4-Bromo-2-ethoxy-1-(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₉H₈BrF₃O₂
Storage Temperature: Ambient
CAS Number: 1355247-61-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR