5-(3-Chloro-5-(trifluoromethoxy)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

59555-1G 59555-500MG 59555-5G
76935-020EA 560.16 USD
76935-020 76935-022 76935-024
5-(3-Chloro-5-(trifluoromethoxy)phenyl)thiazol-2-amine ≥97%
5-(3-Chloro-5-(trifluoromethoxy)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₆ClF₃N₂OS
MW: 294.681 g/mol
Storage Temperature: Ambient
CAS Number: 2755719-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR