5-(4-Ethoxy-2-methylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

59551-1G 59551-500MG 59551-5G
76935-006EA 560.16 USD
76935-006 76935-008 76935-010
5-(4-Ethoxy-2-methylphenyl)thiazol-2-amine ≥97%
5-(4-Ethoxy-2-methylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂OS
MW: 234.317 g/mol
Storage Temperature: Ambient
CAS Number: 2755723-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR