5-(2-Chloro-5-(trifluoromethoxy)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

59548-1G 59548-500MG 59548-5G
76934-994EA 533.64 USD
76934-994 76934-996 76934-998
5-(2-Chloro-5-(trifluoromethoxy)phenyl)thiazol-2-amine ≥97%
5-(2-Chloro-5-(trifluoromethoxy)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₆ClF₃N₂OS
MW: 294.681 g/mol
Storage Temperature: Ambient
CAS Number: 2755723-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR